Learn More
FADS2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA548353
Catalog No. PA548353
Add to cart
Description
Description
Predicted to react with monkey based on sequence homology.
The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13.1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
Specifications
Specifications
FADS2
Polyclonal
Unconjugated
FADS2
Liquid
FADS2
KLH conjugated synthetic peptide between 79-108 amino acids from the N-terminal region of human FADS2.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
O95864
Fatty acid desaturase 2, 11419-, Delta(6) fatty acid desaturase, D6D, Delta(6) desaturase, Delta-6 desaturase, FADS2
Rabbit
IgG
200μl
-20° C, Avoid Freeze/Thaw Cycles
Primary
9415