Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAF1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI10D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271679AF647

Catalog No. N271679A647


Add to cart

Description

Description

FAF1 Monoclonal antibody specifically detects FAF1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FAF1
OTI10D1
Western Blot
Affinity Purified, Conjugated
FAF1
Recombinant protein fragment corresponding to amino acids 415-650 of human FAF1 (NP_008982) produced in E.coli.
Immunogen affinity purified
Apoptosis, Cancer
Primary
11124.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit