Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149281AP

Catalog No. 50-172-9843


Add to cart

Description

Description

This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).
Specifications

Specifications

FAH
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P16930, P25093, P35505
Beta diketonase, FAA, FAH, Fumarylacetoacetase, Fumarylacetoacetate hydrolase
Rabbit
IgG
150 μL
-20°C
Primary
14085, 2184, 29383
Western Blot
Unconjugated
FAH
Liquid
Fah
FAH Fusion Protein Ag6704
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit