Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAHD2A Mouse anti-Human, Clone: OTI9D4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500944

Catalog No. 50-167-2084


Add to cart

Description

Description

May have hydrolase activity.
Specifications

Specifications

FAHD2A
Monoclonal
0.9 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96GK7
CGI-105
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51011
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI9D4
Unconjugated
FAHD2A
Liquid
FAHD2A
Full length human recombit protein of human FAHD2A produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit