Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAHD2B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549012

Catalog No. PA549012


Add to cart

Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

May have hydrolase activity.
Specifications

Specifications

FAHD2B
Polyclonal
PBS with 0.09% sodium azide
Q6P2I3
Fumarylacetoacetate hydrolase domain-containing protein 2B, 3---, FAHD2B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
151313
Western Blot
Unconjugated
FAHD2B
Liquid
FAHD2B
KLH conjugated synthetic peptide between 272-298 amino acids from the C-terminal region of human FAHD2B.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit