Learn More
FAIM3 Mouse anti-Human, Alexa Fluor™ 350, Clone: 992338, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9494U100UG
Catalog No. FAB9494U100
Add to cart
This item is not returnable. View return policy
Description
Description
FAIM3 Monoclonal antibody specifically detects FAIM3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
FAIM3
992338
Flow Cytometry 0.25-1 μg/106 cells
O60667
FAIM3, Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, Toso, TOSOIgM Fc receptor
Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession No. O60667
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human Fc mu R/FAIM3 in direct ELISAs. Detects human, mouse, and rat Fc mu R/FAIM3 in Western blots.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
9214.0
Human