Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAK Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI4D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271268AF700

Catalog No. N271268A700


Add to cart

Description

Description

FAK Monoclonal antibody specifically detects FAK in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

FAK
OTI4D11
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
PTK2
Full length recombinant protein of human FAK (NP_722560) produced in HEK293T cells.
Immunogen affinity purified
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
5747.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, FADK 1, FADKFAK-related non-kinase polypeptide, FAK1FRNK, FAKEC 2.7.10.2, focal adhesion kinase 1, pp125FAK, Protein-tyrosine kinase 2, PTK2 protein tyrosine kinase 2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit