Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Falcon™ FluoroBlok™ 24 Well Permeable Supports and Companion Plates

Block light transmission, simplifying detection of migrating fluorescently labeled cells. Corning™ Falcon™ FlouroBlok™ 24 Well Plate Permeable Supports are designed with a unique light-tight PET membrane. Must be used with Falcon™ 24 Well Companion Plate.

Manufacturer:  Corning™ 351150

 View more versions of this product

Catalog No. 08-772-140

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Dyed PET microporous membrane efficiently blocks the transmission of light within the range of 400-700 nm
  • Fluorescently labeled cells present in the top chamber of the insert are made invisible by the Corning™ FluoroBlok™ membrane
  • Migrated cells are no longer shielded from the light source and are easily detected with a fluorescence plate reader or inverted fluorescence microscope
  • Store at room temperature, away from direct sunlight.
Applications
  • Study a wide range of cell types and activities such as: 
  • Inflammation with neutrophils, transepithelial and transendothelial migration; and analysis of blood-brain barrier, dendritic cells, and Macrophages
  • Pathways for stem cell differentiation
  • Screening for population-specific neuronal motogens
  • Migration of normal, transformed and transfected cells
  • Chemoinvasion assays, drug discovery
NOTE: Individual inserts must be used with Falcon Cell Culture Insert Companion plates to properly position the inserts in the wells.  These plates accurately fit the Corning FluoroBlok 24 Multiwell insert. The Corning FluoroBlok 96 Multiwell inserts use the supplied square well receiver plates. These plates must be used for running the assay, labeling and reading samples to achieve reliable assay results.
Description & Specifications

Specifications

Yes
0.25 in.
Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate
Polystyrene
24
Yes
0.3cm2
0.65cm
24-well
Polyethylene Terephthalate
1μm
FluoroBlok™ Cell Culture Inserts for 24 well plates
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit