Learn More
Falcon™ 24-Multiwell FluoroBlok™ Cell Culture Inserts Systems
Collect data without plate washing, cell scraping, and counting; chart cell migration or bioavailability in real time without dismantling or destroying the insert
Manufacturer:  Corning 351155
 View more versions of this product
Catalog No. 08-772-145
Add to cart
Description
Description
  • Homogeneous fluorescence assay system
  • Chart cell migration or bioavailability in real time without dismantling or destroying the insert
  • Each insert is fitted with a fluorescence blocking Corning Fluoroblok membrane
  • Migrated cells below the membrane are easily detected by a fluorescence plate reader while those remaining above the membrane are made “invisible”
Order Info
Available in 3µ and 8µm pore sizes. One-piece 24-Multiwell insert system (insert plate, 24-well plate or single-well feeder tray, and lid) or 24-well individual cell culture inserts.
Specifications
Specifications
0.3cm2
PET
3μm
With Lid
Sterile
Fluorescence plate reader or inverted fluorescence microscope
24
1/Pk.
FluoroBlok™ HTS 24-Well Multiwell Permeable Support System