Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Falcon™ FluoroBlok™96 Well HTS Insert Systems

Simplify detection of migrating fluorescently labeled cells. Corning™ Falcon™ FluoroBlok™96 Well HTS Insert Systems are designed with a unique light-tight PET membrane that blocks the transmission light within the range of 400 to 700nm.

Manufacturer:  Corning 351161

 View more versions of this product

Catalog No. 08-771-004


Add to cart

Description

Description

 

  • Homogeneous fluorescence assay system
  • Collect data without plate washing, cell scraping, and counting
  • Chart cell migration or bioavailability in real time without dismantling or destroying the insert
  • Each insert is fitted with a fluorescence blocking Corning Fluoroblok membrane
  • Migrated cells below the membrane are easily detected by a fluorescence plate reader while those remaining above the membrane are made “invisible”
  • Migrated cells are no longer shielded from the light source and are easily detected with a fluorescence plate reader or inverted fluorescence microscope
  • Store at room temperature, away from direct sunlight.

Applications
  • Study a wide range of cell types and activities such as: 
  • Inflammation with neutrophils, transepithelial and transendothelial migration; and analysis of blood-brain barrier, dendritic cells, and Macrophages
  • Pathways for stem cell differentiation
  • Screening for population-specific neuronal motogens
  • Migration of normal, transformed and transfected cells
  • Chemoinvasion assays, drug discovery
Specifications

Specifications

0.08cm2
Insert plate with 96 square well, flat bottom plate and lid
PET
3μm
With Lid
Sterile
Fluorescence plate reader or inverted fluorescence microscope
Polystyrene
96
1/Cs.
FluoroBlok™ HTS 96-Well Multiwell Permeable Support System
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit