Learn More
Corning™ Falcon™ Cell Culture Multi Flasks Available on GSA/VA Contract for Federal Government customers only.
Grow cells faster and easier, increasing productivity in cell culture workflow. Corning™ Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
Manufacturer:  Corning™ 353143
 View more versions of this product
Catalog No. 14-826-95
  Check Availability
Add to cart
Specifications
Description
  • Mixing cells and reagents in the BD Falcon Multi-Flask saves time and reduces risk of contamination
  • Flexible design; pour or aspirate/recover cells using a pipet 
  • Consistent surface treatment for predictable scale-up
  • Lot number printed for traceability
  • Manufactured in compliance to cGMP standards
  • Same workflow as standard T-175 Flask
Description & Specifications
Specifications
Vented
Straight
Sterile
PS (Polystyrene)
525cm2
Rectangular
3 Layer tissue culture treated