Learn More
Falcon™ Companion Plates for Cell Culture Inserts Available on GSA/VA Contract for Federal Government customers only.
Feature a one-way, “labyrinth” lid design and condensation rings to minimize evaporation and contamination. Corning™ Falcon™ Companion Plates for Cell Culture Inserts are specifically designed for use with Falcon or Corning™ BioCoat™ Cell Culture Inserts.
Manufacturer:  Corning 353502
 View more versions of this product
Catalog No. 08-771-24
Add to cart
This item is not returnable. View return policy
Description
Description
  • Companion plate wells have notches that hold the insert in place, permit pipet dispensing into the plate well, and ensure proper lid fit
  • Sterile, tissue culture treated, nonpyrogenic
  • May be used with or without cell culture inserts
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Clear
Cell culture inserts
Polystyrene
2.7 to 3.2mL
Sterile
9.6cm2
6 well
6
With Lid