Learn More
Falcon™ Disposable Aspirating Pipets Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polystyrene as a safer alternative to glass Pasteur pipets. Corning™ Falcon™ Disposable Aspirating Pipets are ideal for tissue culture usage or any non-toxic solution that requires aliquoted or aspirating/dispensing in a sterile environment.
Manufacturer:  Corning, Inc. 357558
 View more versions of this product
Catalog No. 13-675-16
  Check Availability
Add to cart
Specifications
Description
  • Convenient for removing spent media from tissue culture vessels
  • No need to autoclave and repackage
  • Ungraduated, unplugged,
  • Sterile, nonpyrogenic
  • Individually wrapped in thermoformed paper/plastic wrap
  • Regulatory status: Class I Medical
Description & Specifications
Specifications
Class I Medical
Aspirating
For tissue culture usage or any non-toxic solution that requires aliquot or aspirating, dispensing in a sterile environment
Yes
Yes
2mL
Polystyrene