Learn More
Falcon™ Erlenmeyer Flasks
High performance polycarbonate Erlenmeyer flask for suspension culture and storage
Manufacturer:  Corning 355123
 View more versions of this product
Catalog No. 08-772-252
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Falcon Erlenmeyer flasks are disposable culture and storage vessels for cell biology, microbiology, or molecular biology applications. They are ideal for use on shaker equipment for culturing bacterial, yeast, and mammalian cells as well as for reagent storage.
  • Provide flexibility with a 2-in-1 vented or non-vented leakproof cap
  • Allow uninhibited visualization of sample or media through the optically clear polycarbonate resin, which is lighter and safer than glass
  • Non-pyrogenic, non-cytotoxic, and nuclease-free
  • The wide mouth design of these flasks allows quick and easy fluid additions and removal, as well as providing easy pipet access to the bottom edges
  • The molded-in graduations provide accurate volumetric measurement for media and buffers
  • 0.2µm PTFE membrane allows for sterile air transfer during aerobic applications
  • Ready-to-use, individually packaged, autoclavable, sterile to SAL 10-6
  • Available in flat- or baffled-bottom, which provides increased agitation and aeration
Compliance
USP Class VI
Specifications
Specifications
Baffled
Store at: 15 to 30°C
Polycarbonate
Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor
16.9 oz.
500mL
Yes
Sterile