Falcon™ Erlenmeyer Flasks

High performance polycarbonate Erlenmeyer flask for suspension culture and storage

Manufacturer: Corning, Inc.Supplier Diversity Partner 355127

 View more versions of this product

Catalog No. 08-772-253

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor
    Capacity (English) 33.8 oz.
    Capacity 33.8 oz., 1000mL
    Bottom Shape Baffled
    Capacity (Metric) 1000mL
    Disposable Yes
    Material Polycarbonate
    Storage Requirements 15 to 30°C
    Sterile Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit