Learn More
Falcon™ Erlenmeyer Flasks
High performance polycarbonate Erlenmeyer flask for suspension culture and storage
Manufacturer:  Corning, Inc. 355121
 View more versions of this product
Catalog No. 08-772-256
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Flat
Store at: 15 to 30°C
Falcon™ Erlenmeyer flasks are culture and storage vessels for cell biology, microbiology, or molecular biology applications, ideal for use on shaker equipment for culturing bacterial, yeast and mammalian cells as well as for reagent stor
250mL
8.45 oz.
Yes
Sterile
Polycarbonate