Falcon™ HTS Multiwell Insert System

Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Corning™ Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.

Manufacturer: Corning, Inc. 351182

 View more versions of this product

Catalog No. 08-777

  N/A
Add to Cart

Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Lid Yes,Yes
    Culture Area 0.3cm2
    For Use With (Application) Excellent for differentiation, toxicology, migration and invasion studies
    Format 24-well,24-well
    Includes 24-well insert plate, 24-well plate and lid
    Material Polystyrene,Polystyrene
    Membrane Polyethylene Terephthalate
    No. of Wells 24
    Pore Size 3μm
    Quantity Ea.
    Sterile Yes
    Type HTS Multiwell Insert System,HTS Multiwell Insert System
    Closure Yes
    Color Transparent
    Wells 24

     

    • Available in 1, 3, and 8µm pore sizes
    • Insert tray is removable from the feeder tray by most robotic arms—reducing the risk of contamination
    • 24-well insert tray distributes media uniformly to each well

     

    Additional feeder trays available

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit