Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Falcon™ In Vitro Fertilization Plasticware: Round Plates Available on GSA/VA Contract for Federal Government customers only.

FDA-cleared 510(k) medical devices

Manufacturer:  Corning 353652

 View more versions of this product

Catalog No. 08-772-42


Due to product restrictions, please to purchase or view availability for this product.

Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. During 2013, they will be integrating Discovery Labware brands (ex. Falcon™, Matrigel™, and BioCoatT™) into their growing family of Corning Life Sciences brands. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

Tested for embryotoxicity by culturing mouse embryos in both the labware itself and in media that has been in contact with the labware for 24 hours at 37°C (98.6°F)

  • Sterile, nonpyrogenic, tissue culture-treated polystyrene plates and dishes
  • Labware is “nontoxic” if it supports growth of ≥75% of the embryos to the expanded and/or hatched blastocyst stage
  • Optically clear for optimum viewing during routine observation and manipulation

Standard 60mm dishes

  • Available with or without 2.45cm2 center well Numbered wells and condensation rings on the lid help prevent any cross-contamination between wells Large writing area allows for easy marking and plate identification

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Supplied in peel-open packaging to ensure sterility. Dishes packed 20 per sleeve; plates individually wrapped.

Specifications

Specifications

Sterile
Polystyrene
54.81mm
21.29cm2
Dish
0.77 oz., 23mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit