Learn More
Falcon™ In Vitro Fertilization Plasticware: Round Plates Available on GSA/VA Contract for Federal Government customers only.
FDA-cleared 510(k) medical devices
Manufacturer:  Corning 353653
Exact match for part number 25382194
 View more versions of this product
Catalog No. 08-772-43
Due to product restrictions, please to purchase or view availability for this product.
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. During 2013, they will be integrating Discovery Labware brands (ex. Falcon™, Matrigel™, and BioCoatT™) into their growing family of Corning Life Sciences brands. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
Tested for embryotoxicity by culturing mouse embryos in both the labware itself and in media that has been in contact with the labware for 24 hours at 37°C (98.6°F)
  • Sterile, nonpyrogenic, tissue culture-treated polystyrene plates and dishes
  • Labware is “nontoxic” if it supports growth of ≥75% of the embryos to the expanded and/or hatched blastocyst stage
  • Optically clear for optimum viewing during routine observation and manipulation
Standard 60mm dishes
  • Available with or without 2.45cm2 center well Numbered wells and condensation rings on the lid help prevent any cross-contamination between wells Large writing area allows for easy marking and plate identification
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Supplied in peel-open packaging to ensure sterility. Dishes packed 20 per sleeve; plates individually wrapped.
Specifications
Specifications
Sterile
Polystyrene
54.81mm
2.89cm2
Dish