Learn More
Falcon™ Plates in Ready-Stack Packaging Available on GSA/VA Contract for Federal Government customers only.
Trays are recyclable, stackable, and reusable for transporting, storing, or incubating plates
Manufacturer:  Corning 353936
 View more versions of this product
Catalog No. 08-772-36
Add to cart
Includes: Lids
This item is not returnable. View return policy
Description
Description
  • Bulk packaged in medical-style peel-open trays for convenient, aseptic handling
  • Plates positioned on their sides, permitting labeling before removal
  • Tissue culture-treated
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Packaged sterile, 14 plates per tray
Specifications
Specifications
Clear
96 well
96
With Lid
96-Well Cell Culture Microplate
0.32cm2
Polystyrene
370mL
0.37mL
Sterile