Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Falcon™ Round-Bottom Polystyrene Tubes Available on GSA/VA Contract for Federal Government customers only.

Constructed of optically clear polystyrene for easy viewing of sample. Corning™ Falcon™ Round-Bottom Polystyrene Tubes are widely used and referenced in laboratory protocols.

Manufacturer:  Corning™ 352017

 View more versions of this product

Catalog No. 14-959-8


  Check Availability
Add to cart

Specifications

Description

  • Individual packages use medical-style packaging materials and allow aseptic access Widely referenced in published laboratory procedures and protocols (e.g., Molecular Cloning, 2nd ed., T. Maniatis et al.)
  • Available with heavyweight, durable, dual-position snap cap for easy handling; with screw cap or without cap
  • All sizes supplied sterile; 12 x 75mm size without cap also available nonsterile

Description & Specifications

Specifications

Clear
17mm
Bag of 125, 1000/Cs.
14mL
100mm
Test Tube
No
1400 x G
Yes
14mL
Without Cap
Polystyrene
Round Bottom
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit