Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Falcon™ Serological Pipets, Bulk Pack Available on GSA/VA Contract for Federal Government customers only.

Color-coded pipets facilitate correct size selection. Corning™ Falcon™ Serological Pipets are ideal for tissue culture or any nontoxic solution.

Manufacturer:  Corning™ 357506

 View more versions of this product

Catalog No. 13-675-10AA


  Check Availability
Add to cart

Specifications

Description

 

  • Gamma-irradiated, nonpyrogenic, made of clear polystyrene for optical clarity
  • Feature exceptionally crisp, dark, easy-to-read printing with both reverse (except on 1mL) and negative graduations
  • Exclusive graduation alignment mark allows graduations to be brought into line of sight before complete removal of the wrapper
  • Inside surface discourages formation of meniscus, thereby reducing reading errors
  • Total delivery accuracy of pipets, ±3%, with a 1.5% coefficient of variation
  • Negative graduations allow for extra capacity
  • Polyester fiber plug helps prevent overfill

 

Description & Specifications

Specifications

1mL
Yellow
Bulk
Standard
±3%
Yes
Polystyrene
Yes
0.4mL
0.01mL
500
1.5%
For tissue culture usage or any non toxic solution
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit