Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FALDH Antibody (OTI2A7), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270159PEATT594
Description
FALDH Monoclonal specifically detects FALDH in Human, Rat samples. It is validated for Flow Cytometry.Specifications
FALDH | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
ALDH3A2 | |
Full length recombinant protein of human FALDH (NP_001026976) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2b |
Flow Cytometry | |
OTI2A7 | |
Flow Cytometry | |
Aldehyde dehydrogenase 10, aldehyde dehydrogenase 3 family, member A2, Aldehyde dehydrogenase family 3 member A2, ALDH10FLJ20851, EC 1.2.1, EC 1.2.1.3, FALDHDKFZp686E23276, fatty aldehyde dehydrogenase, Microsomal aldehyde dehydrogenase, SLS | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
224.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction