Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FALDH Mouse anti-Human, PE/Cy5.5, Clone: OTI2A7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270159PECY55

Catalog No. N270159PEC5


Add to cart

Description

Description

FALDH Monoclonal antibody specifically detects FALDH in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

FALDH
OTI2A7
Flow Cytometry
Affinity Purified, Conjugated
ALDH3A2
Full length recombinant protein of human FALDH (NP_001026976) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
224.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Aldehyde dehydrogenase 10, aldehyde dehydrogenase 3 family, member A2, Aldehyde dehydrogenase family 3 member A2, ALDH10FLJ20851, EC 1.2.1, EC 1.2.1.3, FALDHDKFZp686E23276, fatty aldehyde dehydrogenase, Microsomal aldehyde dehydrogenase, SLS
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit