Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM101A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551321

Catalog No. PA551321


Add to cart

Description

Description

The antibody detects endogenous levels of total FAM101A protein.

Involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in actin cytoskeleton formation in developing cartilaginous cells.
Specifications

Specifications

FAM101A
Polyclonal
Unconjugated
ZNF664-RFLNA
Liquid
ZNF664-RFLNA
Fusion protein of human FAM101A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6ZTI6
CFM2
Rabbit
IgG
100 μL
-20°C
Primary
100533183
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit