Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM101A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | FAM101A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17952820
|
Novus Biologicals
NBP17952820UL |
20 μL |
Each for $152.22
|
|
NBP179528
|
Novus Biologicals
NBP179528 |
100 μL |
Each for $436.00
|
|
Description
FAM101A Polyclonal specifically detects FAM101A in Human samples. It is validated for Western Blot.Specifications
FAM101A | |
Polyclonal | |
Rabbit | |
EAW98447 | |
144347 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
family with sequence similarity 101, member A, FLJ44614 | |
FAM101A | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title