Learn More
Aviva Systems Biology Corp OTULINL Antibody - middle region (ARP49425_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM105A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM105A. Alternate names: FLJ11127, NET20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60082