Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM105A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561267
Description
Immunogen sequence: MAATRSPTRA RERERSGAPA AGSDQVHSWM LATSQALDT Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 56%.
FAM105A belongs to the FAM105 family. The exact function of FAM105A remains unknown.
Specifications
FAM105A | |
Polyclonal | |
Unconjugated | |
FAM105A | |
9830126M18; FAM105A; family with sequence similarity 105 member A; family with sequence similarity 105, member A; inactive ubiquitin thioesterase FAM105A; Inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase FAM105A; LOW QUALITY PROTEIN: inactive ubiquitin thioesterase FAM105A; NET20; OTU deubiquitinase with linear linkage specificity like; OTULINL; protein FAM105A; Protein FAM105A-like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54491 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUU6 | |
FAM105A | |
Recombinant protein corresponding to Human FAM105A. Recombinant protein control fragment (Product #RP-95648). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction