Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ FAM107B Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP258052PEP
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human FAM107B. Source: E.coli Amino Acid Sequence: ASFNQSGVADTHSTVRVQPVAKAGRQPRHPSAEGAPEKRQDSSTHAERNGSANRNSSHRTAAQPAETPEDVPGSLDDG The FAM107B Recombinant Protein Antigen is derived from E. coli. The FAM107B Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
83641 | |
FAM107B Recombinant Protein Antigen | |
PBS and 1M Urea, pH 7.4. | |
C10orf45, chromosome 10 open reading frame 45, family with sequence similarity 107, member B, FLJ45505, hypothetical protein LOC83641, MGC11034, MGC90261 | |
Unlabeled | |
100μL | |
E.Coli |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at −20C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
FAM107B | |
Recombinant Protein Antigen | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only