Learn More
Aviva Systems Biology Corp FAM108B1 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP56805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM108B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM108B1. Alternate names: C9orf77, CGI-67, RP11-409O11.2, FAM108B1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61995
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.