Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD17C Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548160

Catalog No. PA548160

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and chicken based on sequence homology.

Specifications

Specifications

ABHD17C
Polyclonal
PBS with 0.09% sodium azide
Q6PCB6, Q8VCV1
Alpha/beta hydrolase domain-containing protein 17C, 3---, ABHD17C, FAM108C1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
58489, 70178
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ABHD17C
Liquid
ABHD17C
KLH conjugated synthetic peptide between 92-121 amino acids from the Central region of human FAM108C1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit