Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM109A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548339

Catalog No. PA548339


Add to cart

Description

Description

Plays a role in endocytic trafficking. Required for receptor recycling from endosomes, both to the trans-Golgi network and the plasma membrane.
Specifications

Specifications

FAM109A
Polyclonal
PBS with 0.09% sodium azide
Q8N4B1
Sesquipedalian-1, Ses1, 27 kDa inositol polyphosphate phosphatase-interacting protein A, IPIP27A, FAM109A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
144717
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FAM109A
Liquid
FAM109A
KLH conjugated synthetic peptide between 3-31 amino acids from the N-terminal region of human FAM109A.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit