Learn More
FAM111B Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205521AP
Catalog No. 50-173-2957
Add to cart
Description
Description
The antibody is specific to FAM111B. This protein has isoforms with MW 135 kDa (EAW73833, 1200 aa), 85 kDa, and 81 kDa.
FAM111B is a 734 amino acid protein that is encoded by a gene that maps to human chromosome 11, which makes up around 4% of human genomic DNA and isconsidered a gene and disease association dense chromosome.The chromosome 11 encoded Atmgene isimportant for regulation ofcellcycle arrest and apoptosisfollowing double strand DNA breaks. Atmmutation leads to the disorderknown as ataxia-telangiectasia. The blood disorders Sickle cell anemia and beta thalassemia are caused by HBB genemutations. Wilms ft. tumors, WAGR syndrome and Denys-Drash syndrome are associatedwithmutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associatedwith defectsin chromosome 11.
Specifications
Specifications
FAM111B
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6SJ93
CANP, FAM111B, Protein FAM111B
Rabbit
IgG
150 μL
-20°C
Primary
374393
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FAM111B
Liquid
FAM111B
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human