Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM116A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564428
Description
Immunogen sequence: QVKVKKLKNL KTLDSKPGVY TSYKPYLNRD EEIIKQLQKG VQQKRPSEAQ SVILRRYF Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.
Guanine nucleotide exchange factor (GEF) for RAB14. Component of an endocytic recycling pathway that is required for the control of ADAM10 transport, shedding of N-cadherin/CDH2 by ADAM9 or ADAM10 and regulation of cell-cell junctions. Required for RAB14 recruitment to recycling endosomes.
Specifications
| FAM116A | |
| Polyclonal | |
| Unconjugated | |
| DENND6A | |
| AFI1A; DENN domain containing 6A; DENN domain-containing protein 6A; DENN/MADD domain containing 6A; DENND6A; FAM116A; family with sequence similarity 116, member A; protein DENND6A; protein FAM116A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 201627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IWF6 | |
| DENND6A | |
| Recombinant protein corresponding to Human FAM116A. Recombinant protein control fragment (Product #RP-103841). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction