Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM116A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | FAM116A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156753
|
Novus Biologicals
NBP156753 |
100 μL |
Each of 1 for $436.00
|
|
Description
FAM116A Polyclonal specifically detects FAM116A in Human samples. It is validated for Western Blot.Specifications
FAM116A | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
family with sequence similarity 116, member A, FLJ34969, hypothetical protein LOC201627 | |
DENND6A | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q8IWF6 | |
201627 | |
Synthetic peptides corresponding to FAM116A(family with sequence similarity 116, member A) The peptide sequence was selected from the middle region of FAM116A. Peptide sequence KPGVYTSYKPYLNRDEEIIKQLQKGVQQKRPSEAQSVILRRYFLELTQSF. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title