Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM117B Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FAM117B
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095184
View Documents
Novus Biologicals
NBP304928100UL
100 μg Each for $320.71
Add to cart
 
NB095185
View Documents
Novus Biologicals
NBP30492820UL
20 μg Each for $138.37
Add to cart
 
Description

Description

FAM117B Polyclonal antibody specifically detects FAM117B in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

FAM117B
Unconjugated
Rabbit
Human, Mouse
ALS2CR13, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 13, DKFZp686H01244, family with sequence similarity 117, member B, FLJ38771, hypothetical protein LOC150864
Recombinant fusion protein containing a sequence corresponding to amino acids 293-372 of human FAM117B (NP_775782.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
150864
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit