Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM118B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548248

Catalog No. PA548248

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, monkey, mouse and rat based on sequence homology.

May play a role in Cajal bodies formation.
Specifications

Specifications

FAM118B
Polyclonal
PBS with 0.09% sodium azide
Q9BPY3
Protein FAM118B, FAM118B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79607
Western Blot
Unconjugated
FAM118B
Liquid
FAM118B
KLH conjugated synthetic peptide between 212-240 amino acids from the Central region of human FAM118B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit