Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM126A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $378.35

Specifications

Antigen FAM126A
Immunogen Synthetic peptide directed towards the C terminal of human FAM126AThe immunogen for this antibody is FAM126A. Peptide sequence MEISEVDEGFYSRAASSTSQSGLSNSSHNCSNKPSIGKNHRRSGGSKTGG.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17980020 View Documents Novus Biologicals
NBP17980020UL
20ul Each for $152.22
Add to cart
 
NBP179800 View Documents Novus Biologicals
NBP179800
100 ul Each for $378.35
Add to cart
 
Description

Description

FAM126A Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

FAM126A
Rabbit
Polyclonal
Unconjugated
84668
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the C terminal of human FAM126AThe immunogen for this antibody is FAM126A. Peptide sequence MEISEVDEGFYSRAASSTSQSGLSNSSHNCSNKPSIGKNHRRSGGSKTGG.
Primary
IgG
Down-regulated by CTNNB1 protein A, DRCTNNB1Aa, family with sequence similarity 126, member A, HLD5, hyccin, Protein FAM126A
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit