Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM130A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP287405

Catalog No. NB035027


Add to cart

Description

Description

FAM130A1 Polyclonal specifically detects FAM130A1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FAM130A1
Unconjugated
PBS, 2% Sucrose
C12ORF2, C12orf22, CSRNP-2, cysteine/serine-rich nuclear protein 2, cysteine-serine-rich nuclear protein 2, FAM130A1, family with sequence similarity 130, member A1, FLJ25576, Protein FAM130A1, TAIP12, TAIP-12chromosome 12 open reading frame 22, TGF-beta-induced apoptosis protein 12
The immunogen is a synthetic peptide directed towards the C-terminal region of Human FAM130A1
100 μg
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Immunogen affinity purified
RUO
Primary
81566
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit