Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM131C Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $349.40

Specifications

Antigen FAM131C
Immunogen Synthetic peptide directed towards the middle region of human FAM131CThe immunogen for this antibody is FAM131C. Peptide sequence RGRVAHLIEWKGWSAQPAGWELSPAEDEHYCCLPDELREARFAAGVAEQF.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17960720
View Documents
Novus Biologicals
NBP17960720UL
20ul
Each for $152.22
Add to cart
 
NBP179607
View Documents
Novus Biologicals
NBP179607
100 ul
Each for $349.40
Add to cart
 
Description

Description

FAM131C Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

FAM131C
Rabbit
Polyclonal
Unconjugated
348487
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the middle region of human FAM131CThe immunogen for this antibody is FAM131C. Peptide sequence RGRVAHLIEWKGWSAQPAGWELSPAEDEHYCCLPDELREARFAAGVAEQF.
Primary
IgG
C1orf117, family with sequence similarity 131, member C, FLJ36766, hypothetical protein LOC348487, RP11-5P18.9
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit