Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM134B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA564943

Catalog No. PIPA564943


Add to cart

Description

Description

Immunogen sequence: ENGMGTNDEDE LSLGLPTELK RKKEQLDSGH RPSKETQSAA GLTLPLNSDQ TFHLMSNLAG DVITAAVTAA IKDQLEGVQQ ALSQAAPIPE EDTDTEEGDD FELLDQSELD QIESELGLTQ DQEAEAQQNK KSSG Highest antigen sequence identity to the following orthologs - mouse 86%, rat 84%.

The function remains unknown.
Specifications

Specifications

FAM134B
Polyclonal
Unconjugated
RETREG1
Liquid
RETREG1
Recombit Human FAM134B
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H6L5
FAM134B; JK-1; JK1; reticulophagy receptor FAM134B; family with sequence similarity 134 member B; protein FAM134B
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
54463
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only