Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM155B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549013

Catalog No. PA549013

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a product belonging to a family of proteins with unknown function. The presence of two transmembrane domains suggests that this protein is a multi-pass membrane protein. [provided by RefSeq, Sep 2011]
Specifications

Specifications

FAM155B
Polyclonal
PBS with 0.09% sodium azide
O75949
Transmembrane protein FAM155B, Protein TED, Transmembrane protein 28, FAM155B, TED, TMEM28
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27112
Western Blot
Unconjugated
FAM155B
Liquid
FAM155B
KLH conjugated synthetic peptide between 358-384 amino acids from the C-terminal region of human FAM155B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit