Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM156A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | FAM156A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180459
|
Novus Biologicals
NBP180459 |
100 μL |
Each of 1 for $436.00
|
|
Description
FAM156A Polyclonal specifically detects FAM156A in Human samples. It is validated for Western Blot.Specifications
FAM156A | |
Polyclonal | |
Rabbit | |
Human | |
family with sequence similarity 156, member A, TMEM29 | |
FAM156A | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_054857 | |
29057 | |
Synthetic peptide directed towards the middle region of human FAM156A. Peptide sequence NRAPHPSSWETLVQGLSGLTLSLGTNQPGPLPEAALQPQETEEKRQRERQ. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title