Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM160B1 Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157822
Description
FAM160B1 Polyclonal specifically detects FAM160B1 in Human samples. It is validated for Western Blot.Specifications
FAM160B1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
A0JPG1 | |
FAM160B1 | |
Synthetic peptides corresponding to FAM160B1 (family with sequence similarity 160, member B1) Antibody(against the N terminal of FAM160B1. Peptide sequence HYYIETSDDKAPVTDTNIPSHLEQMLDILVQEENERESGETGPCMEYLLH. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%. | |
Human, Mouse, Rat, Amphibian, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
DKFZp686D10123, family with sequence similarity 160, member B1, hypothetical protein LOC57700, KIAA1600bA106M7.3 | |
Rabbit | |
Affinity Purified | |
RUO | |
57700 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title