Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM180A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554354
Description
Immunogen sequence: MCHRWSRAVL FPAAHRPKRS SSLPLNPVLQ TSLEEVELLY EFLLAELEIS PDLQISIKDE ELASLRKASD FRTVCNN Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

Specifications
FAM180A | |
Polyclonal | |
Unconjugated | |
FAM180A | |
FAM180A; family with sequence similarity 180 member A; family with sequence similarity 180, member A; HWKM1940; protein FAM180A; UNQ1940; UNQ1940/PRO4423 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWF9 | |
FAM180A | |
Recombinant protein corresponding to Human FAM180A. Recombinant protein control fragment (Product #RP-92839). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction