Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM183A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583953
Description
Immunogen sequence: PMSWHDNLEE PADARFLNLI HHAAQGPTKK YPE.
FAM183A is a protein coding gene. Gene ontology (GO) annotation include ciliary base.
Specifications
FAM183A | |
Polyclonal | |
Unconjugated | |
FAM183A | |
FAM183A; family with sequence similarity 183 member A; family with sequence similarity 183, member A; hCG23177; LOC440585; protein FAM183A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
440585 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NL82 | |
FAM183A | |
Recombinant protein corresponding to Human FAM183A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction