Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM184B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560616
Description
Immunogen sequence: ARQENSELKG TAKKLGEKLA VAKDRMMLQE CRGTQQTDAM KTELVSENKV LREENDLEAG NLHPQQDQSC LKECPCMKGG TDMQTKKEAS AETEYMKQQY EEDL Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 60%.
KIAA1276 was first identified as a hypothetical protein predicted from the in silico analysis of long cDNAs isolated in the Kazusa cDNA sequencing project. The function of KIAA1276 has not been characterized; however KIAA1276 homozygous knockout mice are viable.
Specifications
FAM184B | |
Polyclonal | |
Unconjugated | |
FAM184B | |
FAM184B; family with sequence similarity 184 member B; family with sequence similarity 184, member B; KIAA1276; Protein FAM184B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULE4 | |
FAM184B | |
Recombinant protein corresponding to Human FAM184B. Recombinant protein control fragment (Product #RP-100189). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction