Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM198B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571113
Description
This target displays homology in the following species: Cow: 100%; Dog: 86%; Guinea Pig: 91%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
FAM198B is a protein coding gene.
Specifications
FAM198B | |
Polyclonal | |
Unconjugated | |
FAM198B | |
1110032E23Rik; 2210419I08Rik; AD021; AD036; AV011458; C4orf18; Ened; Expressed in nerve and epithelium during development; FAM198B; family with sequence similarity 198 member B; family with sequence similarity 198, member B; gask1b; GaskbB; golgi associated kinase 1B; Golgi-associated kinase 1B; LOW QUALITY PROTEIN: protein FAM198B; protein ENED; protein ENED; protein FAM198B; protein FAM198B; UNQ2512/PRO6001 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51313 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UWH4 | |
FAM198B | |
synthetic peptide directed towards the N-terminal region of human FAM198B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction