Learn More
Aviva Systems Biology Corp FAM20C Antibody - C-terminal region (ARP49490_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49490_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM20C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FAM20C. Alternate names: DMP4, RNS, DMP-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60096