Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM219B Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326092JF549
Description
FAM219B Polyclonal antibody specifically detects FAM219B in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
FAM219B | |
Polyclonal | |
Western Blot, Immunocytochemistry | |
C15orf17, chromosome 15 open reading frame 17, FLJ00005, hypothetical protein LOC57184 | |
Produced in rabbits immunized with E. coli-derived Human FAM219B fragment | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
57184 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction